Order Entry
Export
Orders LinkContactUsLinkComponent
 

Tricine

Glass bottle for solids, 1 kg
Catalog # A1085.1000
Supplier:   PanReac AppliChem
Tricine
Catalog # A1085.1000
Supplier:  PanReac AppliChem
CAS Number:  5704-04-1
Tricine

Glass bottle for solids, 1 kg

CAS Number:  5704-04-1
Price
525,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
  • CAS Number:5704-04-1
  • Formula:(HOCH₂)₃CNHCH₂CO₂H
  • MDL Number:MFCD00004277
  • Molecular Weight:179.17 g/mol
  • Merck Index:13,09724
  • Melting Pt:187 °C
  • Density:1.523 g/cm³ (19.85 °C)
  • Storage Temperature:Ambient
  • EINECS:227-193-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications