Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Trehalose dihydrate

D-(+)-Trehalose dihydrate

Glass bottle for solids, 100 g
Catalog # A1742.0100
Supplier:   PanReac AppliChem
D-(+)-Trehalose dihydrate
Catalog # A1742.0100
Supplier:  PanReac AppliChem
CAS Number:  6138-23-4
D-(+)-Trehalose dihydrate
D-(+)-Trehalose dihydrate

Glass bottle for solids, 100 g

CAS Number:  6138-23-4
Price
291,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate
  • CAS Number:6138-23-4
  • Formula:C₁₂H₂₂O₁₁·2H₂O
  • MDL Number:MFCD00071594
  • Molecular Weight:378.33 g/mol
  • Melting Pt:97…99 °C
  • Storage Temperature:Ambient
  • EINECS:202-739-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications