Order Entry
Export
Orders LinkContactUsLinkComponent
 

Streptomycin sulphate

Glass bottle for solids, 1 kg
Catalog # A1852.1000
Health
Irritant
Supplier:   PanReac AppliChem
Streptomycin sulphate
Catalog # A1852.1000
Supplier:  PanReac AppliChem
CAS Number:  3810-74-0
Streptomycin sulphate

Glass bottle for solids, 1 kg

CAS Number:  3810-74-0
Price
1 070,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:3810-74-0
  • Formula:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
  • MDL Number:MFCD00037023
  • Molecular Weight:1457.4 g/mol
  • Merck Index:12,08983
  • Melting Pt:250 °C
  • Storage Temperature:Refrigerator
  • EINECS:223-286-0
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications