Rubidium chloride
Glass bottle for solids, 50 g
Catalog # A4240.0050
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: Rubidiumchloride
- CAS Number:7791-11-9
- Formula:RbCl
- MDL Number:MFCD00011187
- Molecular Weight:120.92 g/mol
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:715 °C
- Density:2.8 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-240-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



