Order Entry
Export
Orders LinkContactUsLinkComponent
Rubidium chloride

Rubidium chloride

Glass bottle for solids, 10 g
Catalog # A4240.0010
Supplier:   PanReac AppliChem
Rubidium chloride
Catalog # A4240.0010
Supplier:  PanReac AppliChem
CAS Number:  7791-11-9
Rubidium chloride

Glass bottle for solids, 10 g

CAS Number:  7791-11-9
Price
148,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Rubidiumchloride
  • CAS Number:7791-11-9
  • Formula:RbCl
  • MDL Number:MFCD00011187
  • Molecular Weight:120.92 g/mol
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:715 °C
  • Density:2.8 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-240-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications