Pancreatin
Glass bottle for solids, 500 g
Catalog # A0585.0500
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Cat. No.:A0585.0500
- Storage conditions:Refrigerator
- Enzyme name:Pancreatin
- CAS No.:8049-47-6



