Order Entry
Export
Orders LinkContactUsLinkComponent
 

Pancreatin

Glass bottle for solids, 100 g
Catalog # A0585.0100
Supplier:   PanReac AppliChem
Pancreatin
Catalog # A0585.0100
Supplier:  PanReac AppliChem
CAS Number:  
Pancreatin

Glass bottle for solids, 100 g

CAS Number:  
Price
55,20 €
Log in for availability
1 * 100 g
Product Details & Documents

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Cat. No.:
    A0585.0100
  • Storage conditions:
    Refrigerator
  • Enzyme name:
    Pancreatin
  • CAS No.:
    8049-47-6

Specifications