Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sodium pyruvate

Glass bottle for solids, 100 g
Catalog # A1530.0100
Sodium pyruvate
Catalog # A1530.0100
Supplier:  
CAS Number:  113-24-6
Sodium pyruvate

Glass bottle for solids, 100 g

CAS Number:  113-24-6
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Sodium pyruvate p. A. 1 * 1 kg Catalog #A1530.1000

 

Product Details & Documents
Synonyms:  2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
  • CAS Number:113-24-6
  • Formula:CH₃COCOONa
  • MDL Number:MFCD00002586
  • Molecular Weight:110.04 g/mol
  • Melting Pt:220…230 °C
  • Density:1.718 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-024-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications