Order Entry
Export
Orders LinkContactUsLinkComponent
 

Guanidinium hydrochloride

Plastic bottle for solids, 1 kg
Catalog # A1499.1000
Irritant
Supplier:   PanReac AppliChem
Guanidinium hydrochloride
Catalog # A1499.1000
Supplier:  PanReac AppliChem
CAS Number:  50-01-1
Guanidinium hydrochloride
Guanidinium hydrochloride

Plastic bottle for solids, 1 kg

CAS Number:  50-01-1
Price
186,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
  • CAS Number:50-01-1
  • Formula:NH₂C(=NH)NH₂·HCl
  • MDL Number:MFCD00013026
  • Molecular Weight:95.53 g/mol
  • Merck Index:13,04578
  • Melting Pt:185 °C
  • Density:1.354 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-002-3
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications