Leupeptin hemisulphate
Glass bottle for solids, 100 mg
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Cat. No.:A2183.0100
- Molecular formula:C₄₀H₇₈N₁₂O₁₂S
- Protein/peptide name:Leupeptin hemisulphate
- Storage conditions:Freezer
- Protein synonyms:(Ac-Leu-Leu-Arg-al hemisulphate) monohydrate
- MDL No.:MFCD00037012
- CAS No.:103476-89-7



