Leupeptin hemisulphate
Glass bottle for solids, 10 mg
A2183.0010
- Pack type:Glass bottle for solids
- Pk:10 mg
- Cat. No.:A2183.0010
- Molecular formula:C₄₀H₇₈N₁₂O₁₂S
- Protein/peptide name:Leupeptin hemisulphate
- Storage conditions:Freezer
- Protein synonyms:(Ac-Leu-Leu-Arg-al hemisulphate) monohydrate
- MDL No.:MFCD00037012
- CAS No.:103476-89-7



