Hygromycin B in solution
5 mL
Catalog # A2175.0005
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
- CAS Number:31282-04-9
- Formula:C₂₀H₃₇N₃O₁₃
- MDL Number:MFCD06795479
- Molecular Weight:527.5 g/mol
Specifications
Specifications
- Pk:5 mL



