Orders LinkContactUsLinkComponent
Bottle pourers
Bottle pourers
Bottle pourer, PTFE, blue, for GL 38 screw neck bottles
Catalog # COWI028.038
Supplier:   COWIE
Bottle pourers
Catalog # COWI028.038
Supplier:  COWIE
Bottle pourers
Bottle pourers
Bottle pourer, PTFE, blue, for GL 38 screw neck bottles
Price
88,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Manufactured in pure, FDA approved, PTFE.
  • Totally chemically resistant and inert
  • To fit GL45, 38, 32 and 30 screw neck bottles
  • State cap colour required, blue, red or green
Permits direct pouring in a safe and consistent manner.
Specifications
  • Type:
    for GL 38 screw neck bottles
  • Description:
    Bottle pourer, PTFE, blue