0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Closures
Test/Sample Tube Closures
Caps for tubes
Caps for tubes
Polyethylene caps, 301201, 401201, 401204, Natural
DELT308202
:
DELTALAB
Caps for tubes
DELT308202
:
DELTALAB
Caps for tubes
Polyethylene caps, 301201, 401201, 401204, Natural
190,00 €
1 * 2.500 items
-
+
Made from polyethylene.
For tubes:
301201, 401201, 401204
Colour:
Natural
Description:
Polyethylene caps
Seq:
02