Eco caps
Eco cap, with three ports
Catalog # JOURJR-9000-0007
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
The Eco Cap consists of a three-hole ETFE insert with a polyethylene collar.
- Compatible with all standard GL 45 thread bottles
- Also available as bottle kit
There is no threading for fittings on the inside of the tubing ports, the tubing can be easily inserted.
Delivery information: The Eco Cap is supplied with one cap, polypropylene male Luer plug. The kit also includes safety-coated glass bottle with a option of 1 or 2 L.
Specifications
- Description:Eco cap, with three ports
- Seq:01