Orders LinkContactUsLinkComponent
Tray, Rotilabo®
Tray, Rotilabo®
365×270×75 mm
Catalog # ROTH8103.1
Supplier:   Roth Carl
Tray, Rotilabo®
Catalog # ROTH8103.1
Supplier:  Roth Carl
Tray, Rotilabo®
365×270×75 mm
Price
106,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Dishes, made of stainless steel, chrome-nickel plated.
  • With folding handles on two sides
  • Very good temperature resistance
  • Easy to clean
  • Autoclavable
Suitable for food applications.
Specifications
  • W×D×H:
    365×270×75 mm