0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
STORAGE SET 13.4 CU.FT. FREESER UPRIGHT 1 * 1 items
REVC6880-81
:
REVCO TECHNOLOGIES
STORAGE SET 13.4 CU.FT. FREESER UPRIGHT 1 * 1 items
REVC6880-81
:
REVCO TECHNOLOGIES