Order Entry
Export
Orders LinkContactUsLinkComponent
Pierce™ Avidin Agarose

Pierce™ Avidin Agarose

Avidin agarose, 5×5 ml, 1 SET
Catalog # PIER20225
Pierce™ Avidin Agarose
Catalog # PIER20225
CAS Number:  
Pierce™ Avidin Agarose
Pierce™ Avidin Agarose

Avidin agarose, 5×5 ml, 1 SET

CAS Number:  
Price
982,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

Pierce™ Avidin agarose is purified avidin immobilised on crosslinked 6% beaded agarose for the affinity purification of biotinylated macromolecules.

  • Strong, nearly irreversible biotin:avidin interaction
  • Approximate binding capacity >20 µg biotin/ml resin
Specifications

Specifications

  • Pk:
    1 SET
  • Description:
    Avidin agarose, 5×5 ml

Specifications