Orders LinkContactUsLinkComponent
VWR®, Yeast extract
Yeast extract, 1 kg
Catalog # J850-1KG
Supplier:   VWR Chemicals
VWR®, Yeast extract
Catalog # J850-1KG
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Yeast extract
VWR®, Yeast extract
Yeast extract, 1 kg
CAS Number:  
Price
321,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Yeast extract is the water soluble portion of autolysed yeast containing a source of Vitamin B complex.
  • High purity extract
  • Used for supplementing media
Specifications
Specifications
  • Pk:
    1 kg
  • Description:
    Yeast extract
Specifications