VWR®, Tris buffer solution, pH 8 (1 mol/L) for biotechnology
TRIS buffer solution pH 8 (1 mol/l), Plastic bottle, 100 mL
Supplier: VWR Chemicals
VWR®, Tris buffer solution, pH 8 (1 mol/L) for biotechnology
Catalog # E199-100ML
Supplier: VWR Chemicals
CAS Number:
Jump to:
Product Details & Documents
Tris (hydroxymethyl) aminomethane.
- Biotechnology grade
- pH @25 °C: 7,9 to 8,1
Storage: room temperature
Documents
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Description:TRIS buffer solution pH 8 (1 mol/l)



