Order Entry
Export
Orders LinkContactUsLinkComponent
Polysorbate 20, Reagent Grade

Polysorbate 20, Reagent Grade

HDPE bottle, 1 L
Catalog # 0777-1L
Supplier:   VWR Chemicals
Polysorbate 20, Reagent Grade
Catalog # 0777-1L
Supplier:  VWR Chemicals
CAS Number:  9005-64-5
Polysorbate 20, Reagent Grade
Polysorbate 20, Reagent Grade

HDPE bottle, 1 L

CAS Number:  9005-64-5
Price
101,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Non-ionic detergent for solubilising membrane proteins without altering biological activity.

Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C
  • Density:1.1 g/cm³ (20 °C)
  • Flash Pt:149 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specification Test Results

  • Arsenic:
    <0.0003%
  • DNase:
    NONE
  • Heavy Metals:
    <0.001%
  • Hydroxyl Number:
    96 – 108
  • Identification:
    PASS
  • Moisture (KF):
    3.0%
  • Protease:
    NONE
  • Residue on Ignition:
    0.25%
  • RNase:
    NONE

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    1 L

Specifications