Silver nitrate ≥99% ACS
Glass bottle for solids, 100 g
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Staining protein after electrophoretic separation. Also used to determine chloride ions in solution.
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- CAS Number:7761-88-8
- Formula:AgNO₃
- MDL Number:MFCD00003414
- Molecular Weight:169.87 g/mol
- Merck Index:12,08661
- Boiling Pt:444 °C (1013 hPa)
- Melting Pt:210 °C
- Density:4.35 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-853-9
- UN:1493
- ADR:5.1,II
- REACH:01-2119513705-43
Documents
Specifications
Specification Test Results
- Appearance (ACS):Colorless or white solid.
- Chloride:≤0.0005%
- Clarity of Solution:Pass
- Copper:≤0.0002%
- Free Acid Test:Pass
- Iron:≤0.0002%
- Lead:≤0.001%
- Purity:≤99%
- Substances not Precipitated by HCl:≤0.01%
- Sulphate:≤0.002%
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



