Order Entry
Export
Orders LinkContactUsLinkComponent
Formamide ≥99%, high purity

Formamide ≥99%, high purity

Plastic bottle, 950 mL
Catalog # 0314-950ML
Health
Supplier:   VWR Chemicals
Formamide ≥99%, high purity
Catalog # 0314-950ML
Supplier:  VWR Chemicals
CAS Number:  75-12-7
Formamide ≥99%, high purity
Formamide ≥99%, high purity

Plastic bottle, 950 mL

CAS Number:  75-12-7
Price
127,00 €
Log in for availability
1 * 950 mL
Product Details & Documents

Ideal for denaturing nucleic acids in hybridisation and DNA sequencing procedures.

Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:13,04261
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:2 °C
  • Density:1.1334 g/cm³ (20 °C)
  • Flash Pt:79 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-842-0

Documents

Specifications

Specification Test Results

  • Abs.@280 nm:
    ≤0.1
  • Conductivity:
    ≤350 umhos
  • Copper:
    <0.0001%
  • Freezing Range:
    1 – 3 °C
  • Iron:
    ≤0.0005%
  • Lead:
    <0.0001%
  • Purity:
    >99%
  • Zinc:
    <0.0001%

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    950 mL

Specifications