Formamide ≥99%, high purity
Plastic bottle, 500 mL
Ideal for denaturing nucleic acids in hybridisation and DNA sequencing procedures.
: Carbamaldehyde
- :75-12-7
- :HCONH₂
- :MFCD00007941
- :45.04 g/mol
- :13,04261
- :210 °C (1013 hPa)
- :2 °C
- :1.1334 g/cm³ (20 °C)
- :79 °C
- :Refrigerator
- :200-842-0
- Abs.@280 nm:≤0.1
- Conductivity:≤350 umhos
- Copper:<0.0001%
- Freezing Range:1 – 3 °C
- Iron:≤0.0005%
- Lead:<0.0001%
- Purity:>99%
- Zinc:<0.0001%
- Pack type:Plastic bottle
- Pk:500 mL



