ContactUsLinkComponent
System angiogenesis BC BioCoat for endothelial cell migration. 24-well cell culture insert system with 3,0 µm FluoroBlok PET membran e. 1 * 1 items
Catalog # 734-1020
Supplier:  Corning
System angiogenesis BC BioCoat for endothelial cell migration. 24-well cell culture insert system with 3,0 µm FluoroBlok PET membran e. 1 * 1 items
Catalog # 734-1020
Supplier:  Corning