Acetone ≥99.5%, GPR RECTAPUR®
Plastic drum, 25 L
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Catalog # 20065.470
Supplier: VWR Chemicals
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56.2 °C (1013 hPa)
- Melting Pt:–95.4 °C
- Density:0.792 g/cm³ (20 °C)
- Flash Pt:<–20 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
More
20065_ACETONE GPR RECTAPUR_BSE_REV1
BSE/TSE Certificates
P20_VWRC20065.293_SPEC_EN.pdf P20_VWRC20065.327_SPEC_EN.pdf P20_VWRC20065.362_SPEC_EN.pdf P20_VWRC20065.464_SPEC_EN.pdf P20_VWRC20065.470_SPEC_EN.pdf
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Related substances:Passes test
- Boiling point:56 - 56.7 °C
- Density (20/4):0.79 - 0.793
- Evaporation residue:Max. 20 ppm
- Assay:Min. 99.5 %
- Acidity:Passes test
- Alkalinity:Passes test
- Appearance of solution:Passes test
- Insolubility in water:Passes test
- IR Spectrum:Passes test
- Substances reducing KMnO4 (as O):Max. 2 ppm
- Water:Max. 0.3 %
Specifications
- Pack type:Plastic drum
- Pk:25 L
- Cat. No.:20065.470
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



