Volumetric flasks, class A, blue markings, BLAUBRAND®
50 ml, 14/23, 0,10 ± ml
612-7136
: Brand
Borosilicate glass 3.3, with PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Individual certificate included
* In addition to DIN.
**Wide neck.
: DIN EN ISO 1042
- NS:14/23
- Capacity:50 ml
- Seq:09
- Tolerance:0,10 ± ml
- Cat. No.:612-7136