Orders LinkContactUsLinkComponent
Stoppers for conical ground joints
7/16, Clear glass
  BRND144605
 :   Brand
Stoppers for conical ground joints
  BRND144605
 :  Brand
Stoppers for conical ground joints
Stoppers for conical ground joints
7/16, Clear glass
5,10 €
1 * 1 items
DURAN®. Hollow, with drip tip and hexagonal grip.
 : DIN 12252.
  • NS:
    7/16
  • Type:
    Clear glass
  • Seq:
    01