Stoppers for conical ground joints
12/21, Clear glass
Catalog # BRND144615
Supplier: Brand
Jump to:
Product Details & Documents
DURAN®. Hollow, with drip tip and hexagonal grip.
Certifications: DIN 12252.
Documents
BRND144635_TDS_EN
Datasheet
Specifications
- NS:12/21
- Type:Clear glass
- Seq:03