Orders LinkContactUsLinkComponent
Funnels for conical ground joints
29/32 NS, 80 mm
  221-0166
 :   Brand
Funnels for conical ground joints
  221-0166
 :  Brand
Funnels for conical ground joints
Funnels for conical ground joints
29/32 NS, 80 mm
35,80 €
1 * 5 items
PP, translucent.
  • Ideal for transferring liquids or powders to multiple-neck flasks
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.
  • Funnel O-Ø flat side:
    80 mm
  • Stem for:
    29/32 NS