Order Entry
Export
Orders LinkContactUsLinkComponent
Chlorine standard solution

Chlorine standard solution

Chlorine standard solution, 2 ml PourRite ampules (NIST), 50 - 75 mg/l as Cl₂, 20
Catalog # HACH1426820
Supplier:   Hach
Chlorine standard solution
Catalog # HACH1426820
Supplier:  Hach
CAS Number:  
Chlorine standard solution
Chlorine standard solution

Chlorine standard solution, 2 ml PourRite ampules (NIST), 50 - 75 mg/l as Cl₂, 20

CAS Number:  
Price
81,80 €
Log in for availability
1 * 20 items
Product Details & Documents

For accuracy checks in chlorine determinations. Lot specific concentration displayed on label.

  • Simple accuracy checks
  • Minimise waste
  • Helps ensure reliable result
Specifications

Specifications

  • Concentration:
    50 - 75 mg/l as Cl₂
  • Pk:
    20
  • Description:
    Chlorine standard solution, 2 ml PourRite ampules (NIST)

Specifications