ContactUsLinkComponent
Thermometer, industrial, straight stem, red liquid filled, measuring range:0:160/1C, capillary lens front, yellow backed, milk glas s scale, upper part:235x20mm, lower part: 213x8mm, cork-glass-top finish. 1 * 1 items
Catalog # AMART90334
Supplier:  Amarell
Thermometer, industrial, straight stem, red liquid filled, measuring range:0:160/1C, capillary lens front, yellow backed, milk glas s scale, upper part:235x20mm, lower part: 213x8mm, cork-glass-top finish. 1 * 1 items
Catalog # AMART90334
Supplier:  Amarell