Lids for Nunc™ microplates
Lid, PS, with condensation rings and evaporation barrier, for use with LowBot™ 384- and 1536-well plates, Clear, 4,6 mm, Non sterile
Catalog # NUNC253623
Supplier: Thermo Fisher Scientific
Lids for Nunc™ microplates
Catalog # NUNC253623
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Documents
Specifications
- Sterility:Non sterile
- Colour:Clear
- Description:Lid, PS, with condensation rings and evaporation barrier, for use with LowBot™ 384- and 1536-well plates
- Height:4,6 mm
- Seq:02
- Cat. No.:NUNC253623
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact