Order Entry
Export
Orders LinkContactUsLinkComponent
 

Diagnostic vials, Nalgene®

10,00 ml, 24×54 mm, Yes, 20-415
  215-1167
   Low Manufacturing Impact
Diagnostic vials, Nalgene®
  215-1167
   Low Manufacturing Impact
Diagnostic vials, Nalgene®
Diagnostic vials, Nalgene®

10,00 ml, 24×54 mm, Yes, 20-415

 
1 220,00 €
   
1 * 500 items
 

These clear PETG vials with white HDPE closures are ideal for sampling, storage and shipment of reagents or buffer solutions.

  • Non-pyrogenic and non-cytotoxic
  • Includes lined, white HDPE closure for a leakproof seal

 : Manufactured in compliance with ISO 13485:2003 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.

 : Packed in shrink-wrapped trays with closures attached. These vials are bulk packed, 100 vials per tray, 500 vials per case.

 
  • Cap included:
    Yes
  • Cap Size:
    20-415
  • Ø×H:
    24×54 mm
  • Capacity:
    10,00 ml
  • Seq:
    02
  • Cat. No.:
    215-1167
  • Environmental certification:
    NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact