Diagnostic vials, Nalgene®
5,00 ml, 22×44 mm, Yes, 20-415
Supplier: Thermo Fisher Scientific
Diagnostic vials, Nalgene®
Catalog # 215-1166
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These clear PETG vials with white HDPE closures are ideal for sampling, storage and shipment of reagents or buffer solutions.
- Non-pyrogenic and non-cytotoxic
- Includes lined, white HDPE closure for a leakproof seal
Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Packed in shrink-wrapped trays with closures attached. These vials are bulk packed, 100 vials per tray, 500 vials per case.
Documents
Specifications
- Cap included:Yes
- Cap Size:20-415
- Ø×H:22×44 mm
- Capacity:5,00 ml
- Seq:01
- Cat. No.:215-1166
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact



