Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
ACCESSORIES FOR SEQUENCE CHAIN 1 * 1 Each
ACCESSORIES FOR SEQUENCE CHAIN 1 * 1 Each
Print
ACCESSORIES FOR SEQUENCE CHAIN 1 * 1 Each
Catalog # WERN1111-003
Supplier:
ENVIROFALK PHARMAWATERSYSTEMS
ACCESSORIES FOR SEQUENCE CHAIN 1 * 1 Each
Catalog # WERN1111-003
Supplier:
ENVIROFALK PHARMAWATERSYSTEMS
Expand image
Return to Top
ACCESSORIES FOR SEQUENCE CHAIN 1 * 1 Each
Add to cart
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 Each
-
+
Inquire For Price
Return Policy
Product Details & Documents