Order Entry
Export
Orders LinkContactUsLinkComponent
Containers, with screw cap, LaboPlast®

Containers, with screw cap, LaboPlast®

250 ml, 68 mm, 79 mm
Catalog # BURK6214-0250
Supplier:   BURKLE
Containers, with screw cap, LaboPlast®
Catalog # BURK6214-0250
Supplier:  BURKLE
Containers, with screw cap, LaboPlast®
Containers, with screw cap, LaboPlast®

250 ml, 68 mm, 79 mm

Price
1,70 €
Log in for availability
1 * 1 items
Product Details & Documents

PP, white.

  • Sterilisable
  • Material impermeable to light
  • Suitable for foodstguffs

Shapely packing container with screw cap. Emptying without residues because there are no crevices.

Specifications
  • Ext.Ø:
    68 mm
  • Capacity:
    250 ml
  • Height:
    79 mm