Order Entry
Export
Orders LinkContactUsLinkComponent
Sub-surface samplers, Wheaton® Grab Sampler® II

Sub-surface samplers, Wheaton® Grab Sampler® II

Grab Sampler® II with sample bottle and PTFE-lined cap, 1000 ml, 549 cm
Catalog # WHEA990450
Supplier:   DWK Life Sciences
Sub-surface samplers, Wheaton® Grab Sampler® II
Catalog # WHEA990450
Supplier:  DWK Life Sciences
Sub-surface samplers, Wheaton® Grab Sampler® II
Sub-surface samplers, Wheaton® Grab Sampler® II

Grab Sampler® II with sample bottle and PTFE-lined cap, 1000 ml, 549 cm

Price
2 370,00 €
Log in for availability
1 * 1 items
Product Details & Documents

The high density PP head holds a narrow mouth borosilicate jar.

  • Remote valve that can be opened by pulling and releasing the finger ring on the handle
  • Aluminum sections can be broken down for easy transport
  • Three lengths available

Certifications: EPA requirements for water and waste water sampling.

Specifications
  • Capacity:
    1000 ml
  • Description:
    Grab Sampler® II with sample bottle and PTFE-lined cap
  • Length:
    549 cm
  • Seq:
    03
Accessories

Product Information

  • Catalog #
    Capacity
    Thread
    Availability
    Price
  • WHEA990477
    1000 ml
    38-430
    Log in for availability
    1 * 1 items - 98,50 €

Product Information

  • Catalog #
    Thread
    Colour
    Availability
    Price
  • Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner
    Screw cap, solid top, phenolic, with PTFE faced 14B styrene-butadiene rubber liner
    WHEA240481
    38-430
    Black
    Log in for availability
    1 * 100 items - 631,00 €