Dispenser bottles, Wheaton®
60 ml, LDPE, 92 mm, 39 mm, 20-410
WHEA211194
These translucent dispensing bottles are excellent for mild and strong buffers with a spout to dispense the contents and then could be depressed to seal the bottle.
- Very flexible with high impact strength
- Good chemical resistance
- Ext.Ø:39 mm
- Thread:20-410
- Capacity:60 ml
- Height:92 mm
- Material:LDPE
- Seq:01



