Order Entry
Export
Orders LinkContactUsLinkComponent
Snap caps for sample bottles, WHEATON®

Snap caps for sample bottles, WHEATON®

34 mm, 30 ml
Catalog # WHEA242616
Supplier:   DWK Life Sciences
Snap caps for sample bottles, WHEATON®
Catalog # WHEA242616
Supplier:  DWK Life Sciences
Snap caps for sample bottles, WHEATON®
Snap caps for sample bottles, WHEATON®

34 mm, 30 ml

Price
103,00 €
Log in for availability
1 * 200 items
Product Details & Documents

Natural low density polyethylene.

  • All sizes have tabs for easy removal
  • Good chemical resistance
  • Good temperature resistance

For WHEATON snap cap sample bottles.

Specifications
  • Cap Size:
    34 mm
  • For bottles:
    30 ml