Orders LinkContactUsLinkComponent
Caps for bottles, open top, Microlink®, WHEATON®
Caps for bottles, open top, Microlink®, WHEATON®
Screw cap, solid top, with PP/silicone/PTFE liner, 20-400, White
Catalog # WHEA240250
Supplier:   DWK Life Sciences
Caps for bottles, open top, Microlink®, WHEATON®
Catalog # WHEA240250
Supplier:  DWK Life Sciences
Caps for bottles, open top, Microlink®, WHEATON®
Caps for bottles, open top, Microlink®, WHEATON®
Screw cap, solid top, with PP/silicone/PTFE liner, 20-400, White
Price
1 270,00 €
Log in for availability
1 * 600 items
Product Details & Documents
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
  • PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
  • Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.
Specifications
  • Thread:
    20-400
  • Colour:
    White
  • Description:
    Screw cap, solid top, with PP/silicone/PTFE liner
  • Seq:
    03