Order Entry
Export
Orders LinkContactUsLinkComponent
Batteries Alkaline and Button Batteries

Batteries Alkaline and Button Batteries

Alkaline button cell batteries, V23, 12 V, 28×10 mm
Catalog # 139-9401
Supplier:   MOINEAU
Batteries Alkaline and Button Batteries
Catalog # 139-9401
Supplier:  MOINEAU
Batteries Alkaline and Button Batteries
Batteries Alkaline and Button Batteries

Alkaline button cell batteries, V23, 12 V, 28×10 mm

Price
5,20 €
Log in for availability
1 * 2 items
Product Details & Documents

Batteries alkaline and button batteries.

  • Shelf life is five times longer than saline batteries
  • As they are waterproof, they will not damage your appliances

Documents

Specifications
  • Type:
    V23
  • Voltage:
    12 V
  • Description:
    Alkaline button cell batteries
  • Ø×H:
    28×10 mm