Diagnostic vials, Nalgene®
10,00 ml, 24×56 mm, Yes, 20-415, +
Supplier: Thermo Fisher Scientific
Diagnostic vials, Nalgene®
Catalog # 391-0075
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These clear PETG vials with white HDPE closures are ideal for sampling, storage and shipment of reagents or buffer solutions.
- Non-pyrogenic and non-cytotoxic
- Sterile to 10⁻⁶ SAL
- 20-415 lined white high-density polyethylene closure attached to PETG vial
- Leakproof when used with Nalgene closures
- Narrow mouth
These bottles are constructed from non-pyrogenic and non-cytotoxic polyethylene terephthalate copolymer with lined white HDPE (high-density polyethylene) closures.
Inner neck diameter: 11 mm
Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Serum Vials meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Packed in shrink-wrapped trays with closures attached.
Specifications
- Cap included:Yes
- Cap Size:20-415
- Ø×H:24×56 mm
- Sterile:+
- Capacity:10,00 ml
- Cat. No.:391-0075
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
NAR Certificate of Retirement - Environmentally Preferable:Low Manufacturing Impact



