Volumetric flasks, class A, white markings
50* ml, 14/23, 0,080 ± ml
Catalog # 612-1546
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, amber, with PE stopper, conformity certified (DE-M marking).
- White enamel indelible markings resistant to acids and alkalis
- Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
Certifications: DIN EN ISO 1042
Documents
Certificates
Search by Lot NumberHirschmann brochure EM-Techcolor -volumetric glassware-
Product Literature
Specifications
- NS:14/23
- Capacity:50* ml
- Tolerance:0,080 ± ml