Volumetric flasks, class A, white markings
25 ml, 10/19, 0,040 ± ml
612-1544
Jump to:
DURAN®, borosilicate glass, amber, with PE stopper, conformity certified (DE-M marking).
- White enamel indelible markings resistant to acids and alkalis
- Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
: DIN EN ISO 1042
Specifications
- NS:10/19
- Capacity:25 ml
- Tolerance:0,040 ± ml