Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
10 ml, 7/16, 0,025 ± ml
Catalog # 612-1541
Supplier:   HIRSCHMANN
Volumetric flasks, class A, white markings
Catalog # 612-1541
Supplier:  HIRSCHMANN
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
10 ml, 7/16, 0,025 ± ml
Price
70,90 €
Log in for availability
1 * 2 items
Product Details & Documents
DURAN®, borosilicate glass, amber, with PE stopper, conformity certified (DE-M marking).
  • White enamel indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
Certifications: DIN EN ISO 1042
Specifications
  • NS:
    7/16
  • Capacity:
    10 ml
  • Tolerance:
    0,025 ± ml