Volumetric flasks, class A, white markings
5* ml, 10/19, 0,040 ± ml
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, amber, with PE stopper, conformity certified (DE-M marking).
- White enamel indelible markings resistant to acids and alkalis
- Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
Certifications: DIN EN ISO 1042
Documents
Certificates
Search by Lot NumberHirschmann brochure EM-Techcolor -volumetric glassware-
Product Literature
Specifications
- NS:10/19
- Capacity:5* ml
- Tolerance:0,040 ± ml