Orders LinkContactUsLinkComponent
Trimming knives
Trimming knives
SM 96
Catalog # SWAN4203
Supplier:   SWANN MORTON
Trimming knives
Catalog # SWAN4203
Supplier:  SWANN MORTON
Trimming knives
Trimming knives
SM 96
Price
67,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Heavy duty trimming knifes with fixed or retractable blade.
  • Blades can be exchanged
  • Trimming knife with a smart black holster that clips onto a belt or waistband
Delivery information: The fixed trimming knife is supplied with holster and three SM92 industrial blades. The retractable knife is supplied with two SM92 industrial blades.
Specifications
  • Type:
    SM 96