Trimming knives
SM 96F
Catalog # SWAN4204
Supplier: SWANN MORTON
Jump to:
Product Details & Documents
Heavy duty trimming knifes with fixed or retractable blade.
- Blades can be exchanged
- Trimming knife with a smart black holster that clips onto a belt or waistband
Delivery information: The fixed trimming knife is supplied with holster and three SM92 industrial blades. The retractable knife is supplied with two SM92 industrial blades.
Specifications
- Type:SM 96F