Orders LinkContactUsLinkComponent
ACM craft tools
No. 28, Handle No. 2 and 5, Spare blade, carbon steel, individually packed, 5 blades per blister card
  SWAN9148
ACM craft tools
  SWAN9148
ACM craft tools
ACM craft tools
No. 28, Handle No. 2 and 5, Spare blade, carbon steel, individually packed, 5 blades per blister card
103,00 €
1 * 10 items
The ACM tool set is presented in a resilient blue carry case and contains a comprehensive range of 13 precision ground carbon steel blades manufactured to a high surgical quality.
Also including three handle variations the set can be used for general cutting applications including the trimming of light and medium weight materials, precision trimming, stencilling, scoring and etching as well as corner cuts, deburring, stripping and gasket cutting making this set extremely versatile.
ACM No. 1 handle - for light duty applications
A light duty craft knife designed to assist with smaller cutting projects on soft or lightweight material.
Suitable blades: No. 7 / 10 / 11 / 16 / 17
ACM No. 2 handle - for medium duty applications
Similar to the light weight knife but slightly thicker to provide added strength and stability.
Suitable blades: No. 2 / 18 / 22 / 24 / 28
ACM No. 5 handle - for heavy duty applications
A heavy duty craft knife that is larger and thicker than all the other types of knives.
Suitable blades: No. 2 / 18 / 22 / 24 / 28
 : All the blades and handles are available on an individual basis or as smaller sets.
  • For:
    Handle No. 2 and 5
  • Type:
    No. 28
  • Description:
    Spare blade, carbon steel, individually packed, 5 blades per blister card